1,1,1-Trifluoroacetylacetone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione

T0434-100G T0434-25G
TCT0434-100GEA 203 USD
TCT0434-100G TCT0434-25G
1,1,1-Trifluoroacetylacetone ≥98.0% (by GC)
1,1,1-Trifluoroacetylacetone

CAS Number: 367-57-7
MDL Number: MFCD00000427
Molecular Formula: C5H5F3O2
Molecular Weight: 154.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 101
Flash Point (°C): 26
Specific Gravity (20/20): 1.28

Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 101 °C
Density: 1.28 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00000427
CAS Number: 367-57-7
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=115ace5a8d5a94df35b603679a20fcc8daef0060&vid=546d01c6fbc823213db03b8c9fd1ebb4e7c8dcd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR