o-Tolidine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4,4'-Bi-o-toluidine

T0253-1G T0253-5G
TCT0253-25GEA 49.85 USD
TCT0253-25G TCT0253-1G TCT0253-5G
o-Tolidine ≥98.0% (by GC)
o-Tolidine

CAS Number: 119-93-7 MDL Number: MFCD00014773 Molecular Formula: C14H16N2 Molecular Weight: 212.30 Purity/Analysis Method: 98.0% (GC) Form: Crystal Color: White Boiling point (°C): 300 Melting point (°C): 129 Flash Point (°C): 244


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials Other Product Names / Synonyms: 4,4'-Diamino-3,3'-dimethylbiphenyl, 3,3'-Dimethylbenzidine

Formula: [-C₆H₃(CH₃)-₄-NH₂]₂
MW: 212.29 g/mol
Boiling Pt: 300 °C
Melting Pt: 129 °C
Flash Pt: 244 °C
MDL Number: MFCD00014773
CAS Number: 119-93-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f38119cb6633c2ea79640baa9648bb10b109f541&vid=f38119cb6633c2ea79640baa9648bb10b109f541&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR