2,4-Dinitrophenyl thiocyanate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,4-Dinitrophenylrhodanide, Thiocyanic Acid 2,4-Dinitrophenyl Ester

T0199-1G T0199-25G
TCT0199-1GEA 52 USD
TCT0199-1G TCT0199-25G
2,4-Dinitrophenyl thiocyanate ≥98.0% (by GC)
2,4-Dinitrophenyl thiocyanate

CAS Number: 1594-56-5
MDL Number: MFCD00047702
Molecular Formula: C7H3N3O4S
Molecular Weight: 225.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 139


Other Product Names / Synonyms: 2,4-Dinitrophenylrhodanide, Thiocyanic Acid 2,4-Dinitrophenyl Ester

Formula: C₇H₃N₃O₄S
MW: 225.18 g/mol
Melting Pt: 139 °C
MDL Number: MFCD00047702
CAS Number: 1594-56-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37fd75a5d0dd4a6e7e58c4ac59ea29d7eaed745c&vid=37fd75a5d0dd4a6e7e58c4ac59ea29d7eaed745c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR