Ethylenetetracarbonitrile ≥98.0%
Supplier: TCI America
Synonyms:
Tetracyanoethylene
T0077-25G
T0077-5G
TCT0077-25GEA
223.86
USD
TCT0077-25G
TCT0077-5G
Ethylenetetracarbonitrile ≥98.0%
Ethylenetetracarbonitrile
CAS Number: 670-54-2
MDL Number: MFCD00001850
Molecular Formula: C6N4
Molecular Weight: 128.09
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Boiling point (°C): 125
Melting point (°C): 198
Lambda max.: 328 nm (CHCl3)
Product Categories: Materials Chemistry, Molecular Conductors, Secondary Battery Research Reagents
Other Product Names / Synonyms: Ethylenetetracarbonitrile, TCNE
Formula:
C₆N₄ MW: 128.09 g/mol Boiling Pt: 223 Melting Pt: 198 °C |
MDL Number:
MFCD00001850 CAS Number: 670-54-2 UN: 3439 ADR: 6.1 II Merck Index: 12,09336 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...