Sodium methoxide ≥96.0%

Supplier: TCI America

Synonyms: Sodium methylate, Sodium methanolate

S0485-100G S0485-500G
TCS0485-100GEA 26.87 USD
TCS0485-100G TCS0485-500G
Sodium methoxide ≥96.0%
Sodium methoxide

CAS Number: 124-41-4
MDL Number: MFCD00012179
Molecular Formula: CH3NaO
Molecular Weight: 54.02
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Flash Point (°C): 78

Formula: CH₃NaO
Flash Pt: 78 °C
MDL Number: MFCD00012179
CAS Number: 124-41-4
UN: 1431
ADR: 4.2,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd649e48f6dc6c0f0d0356257723be954da778fd&vid=fd649e48f6dc6c0f0d0356257723be954da778fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR