Silver hexafluoroantimonate ≥97.0%
Supplier: TCI America
Synonyms:
Silver hexafluoroantimonate(V)
S0463-25G
S0463-5G
TCS0463-25GEA
455
USD
TCS0463-25G
TCS0463-5G
Silver hexafluoroantimonate ≥97.0%
Silver hexafluoroantimonate
CAS Number: 26042-64-8
MDL Number: MFCD00003401
Molecular Formula: AgF6Sb
Molecular Weight: 343.62
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Formula:
AgF₆Sb MW: 343.62 |
MDL Number:
MFCD00003401 CAS Number: 26042-64-8 UN: 3260 ADR: 8 III |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...