Bis(2,2,6,6-tetramethyl-4-piperidyl)sebacate ≥98.0%

Supplier: TCI America

Synonyms: Sebacic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl) Ester, Bis(2,2,6,6-tetramethylpiperidin-4-yl) decanedioate, Decanedioic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl) Ester

S0448-250G S0448-25G S0448-5G
TCS0448-250GEA 109.86 USD
TCS0448-250G TCS0448-25G TCS0448-5G
Bis(2,2,6,6-tetramethyl-4-piperidyl)sebacate ≥98.0%
Bis(2,2,6,6-tetramethyl-4-piperidyl)sebacate

CAS Number: 52829-07-9
MDL Number: MFCD00134709
Molecular Formula: C28H52N2O4
Molecular Weight: 480.73
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 83


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Sebacic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl) Ester, Decanedioic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl) Ester

Formula: C₂₈H₅₂N₂O₄
MW: 480.73 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00134709
CAS Number: 52829-07-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55edb289547298e051a130810c3d990d5eab45d4&vid=55edb289547298e051a130810c3d990d5eab45d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR