Silver tetrafluoroborate ≥98.0%
Supplier: TCI America
Synonyms:
Argentous tetrafluoroborate
S0446-25G
S0446-5G
TCS0446-25GEA
298
USD
TCS0446-25G
TCS0446-5G
Silver tetrafluoroborate ≥98.0%
Silver tetrafluoroborate
CAS Number: 14104-20-2
MDL Number: MFCD00003408
Molecular Formula: AgBF4
Molecular Weight: 194.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 73
Formula:
AgBF₄ MW: 194.67 g/mol Melting Pt: 200 |
MDL Number:
MFCD00003408 CAS Number: 14104-20-2 UN: 3260 ADR: 8 II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...