Sodium cyanotrihydroborate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Sodium cyanoborohydride

S0396-250G S0396-25G
TCS0396-250GEA 1174 USD
TCS0396-250G TCS0396-25G
Sodium cyanotrihydroborate ≥95.0% (by titrimetric analysis)
Sodium cyanotrihydroborate

[Reducing Agent]
CAS Number: 25895-60-7
MDL Number: MFCD00003516
Molecular Formula: CH4BN
Molecular Weight: 62.84
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Reduction, Bioscience, Crosslinkers
Other Product Names / Synonyms: Sodium Cyanotrihydridoborate

Formula: CH₃BNNa
MW: 62.84 g/mol
MDL Number: MFCD00003516
CAS Number: 25895-60-7
UN: 3134
ADR: 4.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=706ed4b30201b209b630cc5900dc820e8fa6df34&vid=87c7bb161cd80c16bb8b7df85f3ffee472087203&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR