Succinyl chloride ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Succinyl dichloride
S0110-250G
S0110-25G
TCS0110-250GEA
232
USD
TCS0110-250G
TCS0110-25G
Succinyl chloride ≥95.0% (by titrimetric analysis)
Succinyl chloride
CAS Number: 543-20-4
MDL Number: MFCD00000749
Molecular Formula: C4H4Cl2O2
Molecular Weight: 154.97
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 192
Flash Point (°C): 76
Freezing point (°C): 14
Specific Gravity (20/20): 1.37
Formula:
C₄H₄Cl₂O₂ MW: 154.98 g/mol Boiling Pt: 192 °C Density: 1.37 (20 °C) Flash Pt: 76 °C |
MDL Number:
MFCD00000749 CAS Number: 543-20-4 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...