Ruthenium(III) chloride

Supplier: TCI America

R0074-1G R0074-5G
TCR0074-1GEA 97.65 USD
TCR0074-1G TCR0074-5G
Ruthenium(III) chloride
Ruthenium(III) chloride

CAS Number: 10049-08-8
MDL Number: MFCD00011208
Molecular Formula: Cl3Ru
Molecular Weight: 207.42
Form: Crystal
Color: Black

Formula: Cl₃Ru
MDL Number: MFCD00011208
CAS Number: 10049-08-8
UN: 3260
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85051b4636d3041d6e05c0cd9014d637a015474a&vid=85051b4636d3041d6e05c0cd9014d637a015474a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR