Dodecamolybdophosphoric acid hydrate

Supplier: TCI America
Danger

P1910-100G P1910-500G
TCP1910-100GEA 104.4 USD
TCP1910-100G TCP1910-500G
Dodecamolybdophosphoric acid hydrate
Dodecamolybdophosphoric acid hydrate

CAS Number: 51429-74-4
MDL Number: MFCD00149913
Molecular Formula: H3Mo12O40P
Molecular Weight: 1825.48
Form: Crystal
Color: Pale Yellow
Melting point (°C): 90


Other Product Names / Synonyms: Molybdo(VI)phosphoric Acid

Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
Melting Pt: 90 °C
MDL Number: MFCD00149913
CAS Number: 51429-74-4
UN: 3085
ADR: 8,III
Merck Index: 12,07498
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40d1d77133aaf6f819deea9b9604bb8b5b9bc667&vid=d8ac726ab77d7276f92eed773e2cc7ac20548bd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Molybdenum oxide min. 72.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR