Paliperidone ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 9-Hydroxyrisperidone
CAS Number: 144598-75-4
Molecular Formula: C23H27FN4O3
Molecular Weight: 426.49
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Slightly Pale Yellow
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one
Formula:
C₂₃H₂₇FN₄O₃ MW: 426.49 g/mol |
MDL Number:
MFCD00871802 CAS Number: 144598-75-4 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...