Pentafluorophenyl trifluoroacetate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Trifluoroacetic Acid Pentafluorophenyl Ester, Pentafluorphenyl trifluoracetate

P1894-25G P1894-5G
TCP1894-25GEA 215.5 USD
TCP1894-25G TCP1894-5G
Pentafluorophenyl trifluoroacetate ≥95.0% (by GC)
Pentafluorophenyl trifluoroacetate

CAS Number: 14533-84-7
MDL Number: MFCD00134438
Molecular Formula: C8F8O2
Molecular Weight: 280.07
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 122
Melting point (°C): 4
Flash Point (°C): 52
Specific Gravity (20/20): 1.64


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Fluorinated Building Blocks, Other Fluorinated Group Introducing Agents
Other Product Names / Synonyms: Trifluoroacetic Acid Pentafluorophenyl Ester

Formula: C₈F₈O₂
MW: 280.07 g/mol
Boiling Pt: 122 °C
Melting Pt: 4 °C
Density: 1.64 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00134438
CAS Number: 14533-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d9b54c1b6e344ffd1f9e2ed47de8932c4f7ab55&vid=5d9b54c1b6e344ffd1f9e2ed47de8932c4f7ab55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR