m-Phenylenediamine ≥98.0%

Supplier: TCI America

Synonyms: MPD, 1,3-Diaminobenzene, 1,3-Phenylenediamine

P1892-25G P1892-500G
TCP1892-25GEA 26.46 USD
TCP1892-25G TCP1892-500G
m-Phenylenediamine ≥98.0%
m-Phenylenediamine

CAS Number: 108-45-2
MDL Number: MFCD00007799
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 287
Melting point (°C): 65
Flash Point (°C): 156


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,3-Diaminobenzene

Formula: C₆H₈N₂
Boiling Pt: 287 °C
Melting Pt: 65 °C
Flash Pt: 156 °C
MDL Number: MFCD00007799
CAS Number: 108-45-2
UN: 1673
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d0216b0b14bc9bfa27be7a06f46d1036eba7662&vid=6d0216b0b14bc9bfa27be7a06f46d1036eba7662&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR