2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride, 2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride

P1890-25G P1890-5G
TCP1890-25GEA 86 USD
TCP1890-25G TCP1890-5G
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride ≥98.0%
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride

CAS Number: 70918-74-0
MDL Number: MFCD00729051
Molecular Formula: C13H16N2O3
Molecular Weight: 284.74
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 263


Other Product Names / Synonyms: 1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride

Formula: C₁₃H₁₆N₂O₃
Melting Pt: 263 °C
MDL Number: MFCD00729051
CAS Number: 70918-74-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de65f9219e957e8f2b2c668896368d150d79260c&vid=de65f9219e957e8f2b2c668896368d150d79260c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR