O-Phenylenediamine ≥98.0% (by GC, titration analysis) for biochemical research

Supplier: TCI America

Synonyms: 1,2-Phenylendiamine, 1,2-Phenylenediamine, 1,2-Diaminobenzene, OPD

P1805-1G P1805-5G
TCP1805-1GEA 46 USD
TCP1805-1G TCP1805-5G
O-Phenylenediamine ≥98.0% (by GC, titration analysis) for biochemical research
O-Phenylenediamine

[for Biochemical Research]
CAS Number: 95-54-5
MDL Number: MFCD00007721
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 265
Melting point (°C): 103
Flash Point (°C): 156


Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 1,2-Diaminobenzene, OPD

Formula: C₆H₈N₂
Boiling Pt: 265 °C
Melting Pt: 103 °C
Flash Pt: 156 °C
MDL Number: MFCD00007721
CAS Number: 95-54-5
UN: 1673
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9671463eec5179f22df6a2f41e1a78e37824c165&vid=fd15c67254e26fd3cf9cc0494987e6e45c294bda&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR