(±)-4-Penten-2-ol ≥99.0% (by GC)

Supplier: TCI America

Synonyms: 4-Penten-2-ol, (±)-1-Penten-4-ol, (±)-Allyl methyl carbinol

P1804-25G P1804-5G
TCP1804-25GEA 214 USD
TCP1804-25G TCP1804-5G
(±)-4-Penten-2-ol ≥99.0% (by GC)
(±)-4-Penten-2-ol

CAS Number: 625-31-0
MDL Number: MFCD00004556
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 116
Flash Point (°C): 30
Specific Gravity (20/20): 0.84

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 116 °C
Density: 0.84 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00004556
CAS Number: 625-31-0
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16c3fa7f0e9a9fd804ce5c97d9d3d4332cf9357&vid=e16c3fa7f0e9a9fd804ce5c97d9d3d4332cf9357&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR