trans-2-Pentenal ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Trans-2-pentenal, trans-2-Pentenal, (E)-2-Pentenal

P1790-25G P1790-5G
TCP1790-25GEA 97 USD
TCP1790-25G TCP1790-5G
trans-2-Pentenal ≥95.0% (by GC)
trans-2-Pentenal

CAS Number: 1576-87-0
MDL Number: MFCD00009615
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 81
Flash Point (°C): 22
Specific Gravity (20/20): 0.85
Storage Temperature: 0-10°C

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 81 °C
Density: 0.85 (20 °C)
Flash Pt: 22 °C
MDL Number: MFCD00009615
CAS Number: 1576-87-0
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62c4bc5cc8be66fe47da15ab441e82b923aa9d63&vid=369e9d129fb1830b63218f508aef89e72d6b3310&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR