S-Propyl thioacetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: S-Propyl ethanethioate

P1716-25G P1716-5G
TCP1716-25GEA 124 USD
TCP1716-25G TCP1716-5G
S-Propyl thioacetate ≥98.0% (by GC)
S-Propyl thioacetate

CAS Number: 2307-10-0
MDL Number: MFCD00039937
Molecular Formula: C5H10OS
Molecular Weight: 118.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 137
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Thioacetic Acid S-Propyl Ester

Formula: C₅H₁₀OS
MW: 118.2 g/mol
Boiling Pt: 137 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00039937
CAS Number: 2307-10-0
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a017b72abac502de14c09c0a3ddd1350da302a8&vid=8a017b72abac502de14c09c0a3ddd1350da302a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR