Phenyl trifluoromethyl sulfide ≥97.0%

Supplier: TCI America

Synonyms: Phenyl(trifluoromethyl)sulphane, alpha,alpha,alpha-Trifluorothioanisole, (Trifluoromethylthio)benzene

P1693-25G P1693-5G
TCP1693-25GEA 337 USD
TCP1693-25G TCP1693-5G
Phenyl trifluoromethyl sulfide ≥97.0%
Phenyl trifluoromethyl sulfide

CAS Number: 456-56-4
MDL Number: MFCD00040839
Molecular Formula: C7H5F3S
Molecular Weight: 178.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 142
Flash Point (°C): 37
Specific Gravity (20/20): 1.27


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: (Trifluoromethylthio)benzene, alpha,alpha,alpha-Trifluorothioanisole

Formula: C₇H₅F₃S
Boiling Pt: 142 °C
Density: 1.27 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00040839
CAS Number: 456-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a78eff143c981089e37260d2659a4d1623347dca&vid=a78eff143c981089e37260d2659a4d1623347dca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR