Potassium tetrahydroborate ≥98.0%

Supplier: TCI America

Synonyms: Potassium tetrahydridoborate, Potassium borohydride

P1681-100G P1681-25G
TCP1681-100GEA 67.12 USD
TCP1681-100G TCP1681-25G
Potassium tetrahydroborate ≥98.0%
Potassium tetrahydroborate

CAS Number: 13762-51-1
Molecular Formula: H4BK
Molecular Weight: 53.94
Purity/Analysis Method: >98.0% (V)
Form: Crystal
Color: White
Melting point (°C): 500


Product Categories: Synthetic Organic Chemistry, Reduction, Materials Chemistry, Fuel Cell Research Reagents
Other Product Names / Synonyms: Potassium Tetrahydridoborate, Potassium Tetrahydroborate

Formula: H₄BK
MW: 53.94 g/mol
Melting Pt: 500
MDL Number: MFCD00011396
CAS Number: 13762-51-1
UN: 1870
ADR: 4.3 I
Merck Index: 12,07777
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=714f9a9fb6ac4954510dc2a61a307ac0e7893e04&vid=714f9a9fb6ac4954510dc2a61a307ac0e7893e04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (V)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR