4-Piperidinemethanol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Piperidin-4-ylmethanol

P1653-25G P1653-5G
TCP1653-25GEA 240 USD
TCP1653-25G TCP1653-5G
4-Piperidinemethanol ≥97.0% (by GC, titration analysis)
4-Piperidinemethanol

CAS Number: 6457-49-4
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 135
Melting point (°C): 59
Flash Point (°C): 123


Other Product Names / Synonyms: 4-(Hydroxymethyl)piperidine

Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 135 °C
Melting Pt: 59 °C
Flash Pt: 123 °C
MDL Number: MFCD00174228
CAS Number: 6457-49-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=344521350a3aea8c1b5ed3c6650b7ad75ce75129&vid=344521350a3aea8c1b5ed3c6650b7ad75ce75129&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR