2-Pyridinesulfonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Pyridine-2-sulphonic acid

P1647-25G P1647-5G
TCP1647-25GEA 272 USD
TCP1647-25G TCP1647-5G
2-Pyridinesulfonic acid ≥98.0% (by HPLC, titration analysis)
2-Pyridinesulphonic acid

CAS Number: 15103-48-7
Molecular Formula: C5H5NO3S
Molecular Weight: 159.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 252

Formula: C₅H₅NO₃S
MW: 159.1631 g/mol
Melting Pt: 252 °C
MDL Number: MFCD00234080
CAS Number: 15103-48-7
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6c3665acab631c7b2cb3e49a46319e1769ea7c0&vid=b6c3665acab631c7b2cb3e49a46319e1769ea7c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR