1,4-Phenylene diisocyanate ≥98.0% (by GC)

Supplier: TCI America

P1640-250G P1640-25G
TCP1640-250GEA 288 USD
TCP1640-250G TCP1640-25G
1,4-Phenylene diisocyanate ≥98.0% (by GC)
1,4-Phenylene diisocyanate

CAS Number: 104-49-4
Molecular Formula: C8H4N2O2
Molecular Weight: 160.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 118
Melting point (°C): 96
Flash Point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Benzene 1,4-Diisocyanate

Formula: C₈H₄N₂O₂
Boiling Pt: 118 °C
Melting Pt: 96 °C
Flash Pt: 113 °C
CAS Number: 104-49-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3627ce0058d5b23dffd11362351bf2cb4e660b0b&vid=3627ce0058d5b23dffd11362351bf2cb4e660b0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR