Pentafluoroanilinium trifluoromethanesulfonate ≥98.0%
Supplier: TCI America
Synonyms:
2,3,4,5,6-Pentafluoroaniline trifluoromethanesulphonate, Pentafluoroanilinium Triflate, PFPAT, (Pentafluorophenyl)ammonium Trifluoromethanesulphonate
P1626-1G
P1626-25G
P1626-5G
TCP1626-1GEA
70.19
USD
TCP1626-1G
TCP1626-25G
TCP1626-5G
Pentafluoroanilinium trifluoromethanesulfonate ≥98.0%
Pentafluoroanilinium trifluoromethanesulfonate
CAS Number: 912823-79-1
Molecular Formula: C7H3F8NO3S
Molecular Weight: 333.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: Pentafluoroanilinium Triflate, (Pentafluorophenyl)ammonium Trifluoromethanesulfonate, PFPAT
Formula:
C₇H₃F₈NO₃S MW: 333.15 g/mol |
MDL Number:
MFCD08276416 CAS Number: 912823-79-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...