Phenol ≥99.5% (by GC)

Supplier: TCI America

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

P1610-25G P1610-500G
TCP1610-25GEA 28.61 USD
TCP1610-25G TCP1610-500G
Phenol ≥99.5% (by GC)
Phenol

CAS Number: 108-95-2
Molecular Formula: C6H6O
Molecular Weight: 94.11
Purity/Analysis Method: >99.5% (GC)
Form: Crystal
Color: White
Boiling point (°C): 181
Melting point (°C): 42
Flash Point (°C): 79

Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.7
Melting Pt: 40.8
Flash Pt: 79 °C
MDL Number: MFCD00002143
CAS Number: 108-95-2
UN: 1671
ADR: 6.1 II
Merck Index: 12,07390
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64b9cdadca882b63d679da786774304b0d5dd60f&vid=8b11625f890dd34a30243238c9e1f87f3a2a95f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR