Trimethoxy[3-(phenylamino)propyl]silane ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-[3-(Trimethoxysilyl)propyl]aniline

P1458-250G P1458-25G
TCP1458-250GEA 236 USD
TCP1458-250G TCP1458-25G
Trimethoxy[3-(phenylamino)propyl]silane ≥98.0% (by titrimetric analysis)
Trimethoxy[3-(phenylamino)propyl]silane

CAS Number: 3068-76-6
MDL Number: MFCD00053673
Molecular Formula: C12H21NO3Si
Molecular Weight: 255.39
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 310
Flash Point (°C): 165
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: N-[3-(Trimethoxysilyl)propyl]aniline

Formula: C₁₂H₂₁NO₃Si
MW: 255.39 g/mol
Boiling Pt: 310 °C
Density: 1.07 (20 °C)
Flash Pt: 165 °C
MDL Number: MFCD00053673
CAS Number: 3068-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1bf5aac78e1ba395a57ba4770ae06bbe5c1173b&vid=c1bf5aac78e1ba395a57ba4770ae06bbe5c1173b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR