2,3,4,5,6-Pentafluorotoluene ≥98.0% (by GC)

Supplier: TCI America

P1408-25G P1408-5G
TCP1408-25GEA 249 USD
TCP1408-25G TCP1408-5G
2,3,4,5,6-Pentafluorotoluene ≥98.0% (by GC)
2,3,4,5,6-Pentafluorotoluene

CAS Number: 771-56-2
MDL Number: MFCD00000298
Molecular Formula: C7H3F5
Molecular Weight: 182.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 34
Specific Gravity (20/20): 1.45

Formula: C₇H₃F₅
MW: 182.09 g/mol
Density: 1.45 (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00000298
CAS Number: 771-56-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa448ab1ddf8705019623112c36990ff6b467efd&vid=c99d8a942a93992eb244991e1bc3c24fd331209b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR