(R)-(+)-1,2-Epoxypropane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (R)-(+)-Propene oxide, (R)-(+)-Methyloxirane, (R)-(+)-1,2-Propylene oxide, (R)-(+)-Propylene oxide

P1396-25ML P1396-5ML
TCP1396-25MLEA 206.52 USD
TCP1396-25ML TCP1396-5ML
(R)-(+)-1,2-Epoxypropane ≥98.0% (by GC)
(R)-(+)-1,2-Epoxypropane

CAS Number: 15448-47-2
MDL Number: MFCD00066211
Molecular Formula: C3H6O
Molecular Weight: 58.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 34
Flash Point (°C): -37
Specific Gravity (20/20): 0.83
Specific rotation [a]20/D: 14 deg (neat)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1,2-Epoxypropane

Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 34 °C
Density: 0.83 (20 °C)
Flash Pt: –37 °C
MDL Number: MFCD00066211
CAS Number: 15448-47-2
UN: 1280
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=167f3c0d71e063b2cb9b1703520f5aab0781b769&vid=167f3c0d71e063b2cb9b1703520f5aab0781b769&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR