4-Propylbenzenesulphonyl chloride ≥93.0%

Supplier: TCI America

Synonyms: 4-Propylbenzene-1-sulphonyl chloride

P1366-25G P1366-5G
TCP1366-25GEA 155.9 USD
TCP1366-25G TCP1366-5G
4-Propylbenzenesulphonyl chloride ≥93.0%
4-Propylbenzenesulphonyl chloride

CAS Number: 146949-07-7
MDL Number: MFCD00041508
Molecular Formula: C9H11ClO2S
Molecular Weight: 218.70
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 110
Specific Gravity (20/20): 1.23

Formula: C₉H₁₁ClO₂S
Density: 1.23 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00041508
CAS Number: 63014-04-0
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1484af68a3e3d573f1f3b8e183c4ddcc37ddeff2&vid=1484af68a3e3d573f1f3b8e183c4ddcc37ddeff2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR