D-Phenylalaninol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: D-Penylalaninol, D(+)-2-Amino-3-phenyl-1-propanol

P1289-25G P1289-5G
TCP1289-25GEA 219 USD
TCP1289-25G TCP1289-5G
D-Phenylalaninol ≥98.0% (by titrimetric analysis)
D-Phenylalaninol

CAS Number: 5267-64-1
MDL Number: MFCD00064298
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 122
Melting point (°C): 93
Specific rotation [a]20/D: 23.5 deg (C=1.2, 1mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-2-Amino-3-phenyl-1-propanol

Formula: C₉H₁₃NO
MW: 151.2 g/mol
Boiling Pt: 122 °C
Melting Pt: 93 °C
MDL Number: MFCD00064298
CAS Number: 5267-64-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3503cd3ba732f3940414439ff9dc51594d54e7f4&vid=3503cd3ba732f3940414439ff9dc51594d54e7f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR