1,4-Phenylene diisothiocyanate ≥96.0% (by titrimetric analysis)

Supplier: TCI America

P1279-25G P1279-5G
TCP1279-25GEA 385 USD
TCP1279-25G TCP1279-5G
1,4-Phenylene diisothiocyanate ≥96.0% (by titrimetric analysis)
1,4-Phenylene diisothiocyanate

CAS Number: 4044-65-9
MDL Number: MFCD00004811
Molecular Formula: C8H4N2S2
Molecular Weight: 192.25
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Melting point (°C): 130


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Benzene 1,4-Diisothiocyanate, Bitoscanate, 1,4-Diisothiocyanatobenzene

Formula: C₈H₄N₂S₂
Melting Pt: 130 °C
MDL Number: MFCD00004811
CAS Number: 4044-65-9
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15032bd5ca2885e69a6e638396e803e286e1968e&vid=15032bd5ca2885e69a6e638396e803e286e1968e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR