Pyrene ≥97.0% (by GC)
Supplier: TCI America
P1104-25G
P1104-500G
P1104-100G
TCP1104-25GEA
62.07
USD
TCP1104-25G
TCP1104-500G
TCP1104-100G
Pyrene ≥97.0% (by GC)
Pyrene
CAS Number: 129-00-0
MDL Number: MFCD00004136
Molecular Formula: C16H10
Molecular Weight: 202.26
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 404
Melting point (°C): 150
Flash Point (°C): 199
Lambda max.: 330 nm (EtOH)
Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: Benzo[def]phenanthrene
Formula:
C₁₆H₁₀ MW: 202.26 g/mol Boiling Pt: 404 °C Melting Pt: 150 °C Flash Pt: 199 °C |
MDL Number:
MFCD00004136 CAS Number: 129-00-0 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...