Perfluorobutane-1-sulphonyl fluoride ≥93.0%
Supplier: TCI America
Synonyms:
Perfluoro-1-butanesulphonyl fluoride, Nonafluorobutanesulphonyl fluoride
P1098-250G
P1098-25G
TCP1098-250GEA
438
USD
TCP1098-250G
TCP1098-25G
Perfluorobutane-1-sulphonyl fluoride ≥93.0%
Perfluorobutane-1-sulphonyl fluoride
CAS Number: 375-72-4
MDL Number: MFCD00007422
Molecular Formula: C4F10O2S
Molecular Weight: 302.09
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 65
Specific Gravity (20/20): 1.72
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds
Other Product Names / Synonyms: Nonafluoro-1-butanesulfonyl Fluoride
Formula:
C₄F₁₀O₂S Boiling Pt: 65 °C Density: 1.72 (20 °C) |
MDL Number:
MFCD00007422 CAS Number: 375-72-4 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >93.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...