1,3-Phenylene diisocyanate ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 123-61-5
MDL Number: MFCD00002018
Molecular Formula: C8H4N2O2
Molecular Weight: 160.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 103
Melting point (°C): 50
Flash Point (°C): 110
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Benzene 1,3-Diisocyanate
Formula:
C₈H₄N₂O₂ Boiling Pt: 103 °C Melting Pt: 50 °C Flash Pt: 110 °C |
MDL Number:
MFCD00002018 CAS Number: 123-61-5 UN: 2811 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...