Benzeneselenenyl chloride ≥97.0%

Supplier: TCI America

Synonyms: Phenylselenium chloride, Phenylselenenyl chloride, Chloroselenobenzene, Phenylselenyl chloride

P1091-25G P1091-5G
TCP1091-25GEA 254 USD
TCP1091-25G TCP1091-5G
Benzeneselenenyl chloride ≥97.0%
Benzeneselenenyl chloride

CAS Number: 5707-04-0
MDL Number: MFCD00000478
Molecular Formula: C6H5ClSe
Molecular Weight: 191.52
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Pale Yellow
Boiling point (°C): 95
Melting point (°C): 63
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Selenenylation
Other Product Names / Synonyms: Benzeneselenenyl Chloride

Formula: (C₆H₅)SeCl
MW: 191.52 g/mol
Boiling Pt: 237
Melting Pt: 63 °C
MDL Number: MFCD00000478
CAS Number: 5707-04-0
UN: 2928
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdf1a3f37291c48ae3f981a9a805585e94e47e43&vid=cdf1a3f37291c48ae3f981a9a805585e94e47e43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR