Phosphorus pentoxide - methanesulfonic acid

Supplier: TCI America

P1042-25G P1042-500G
TCP1042-25GEA 30.21 USD
TCP1042-25G TCP1042-500G
Phosphorus pentoxide - methanesulfonic acid
Phosphorus pentoxide - methanesulfonic acid

CAS Number: 39394-84-8
MDL Number: MFCD00070545
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 122
Flash Point (°C): 113


Other Product Names / Synonyms: Eaton's Reagent

Boiling Pt: 122 °C
Flash Pt: 113 °C
MDL Number: MFCD00070545
CAS Number: 39394-84-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af5f4dd5c1e2f1be5af14d7e523d3657d08d1532&vid=af5f4dd5c1e2f1be5af14d7e523d3657d08d1532&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR