Tin(II) phthalocyanine
Supplier: TCI America
Synonyms:
Tin(ii)phthalocyanine
P1024-1G
P1024-5G
TCP1024-1GEA
54
USD
TCP1024-1G
TCP1024-5G
Tin(II) phthalocyanine
Tin (II) phthalocyanine
CAS Number: 15304-57-1
MDL Number: MFCD00058867
Molecular Formula: C32H16N8Sn
Molecular Weight: 631.25
Form: Crystal
Color: Purple
Other Product Names / Synonyms: Phthalocyanine Tin(II)
Formula:
C₃₂H₁₆N₈Sn MW: 631.233 g/mol |
MDL Number:
MFCD00058867 CAS Number: 15304-57-1 UN: 3146 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...