Chloromethyl pivalate ≥99.0% (by GC)

Supplier: TCI America

Synonyms: Chloromethyl trimethylacetate, Trimethylacetic acid chloromethyl ester, 2,2-Dimethylpropionic acid chloromethyl ester, Pivalic acid chloromethyl ester

P1012-25G P1012-500G P1012-100G
TCP1012-25GEA 27 USD
TCP1012-25G TCP1012-500G TCP1012-100G
Chloromethyl pivalate ≥99.0% (by GC)
Chloromethyl pivalate

[Amino-Protecting Agent]
CAS Number: 18997-19-8
MDL Number: MFCD00000884
Molecular Formula: C6H11ClO2
Molecular Weight: 150.60
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 147
Flash Point (°C): 40
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Chloromethyl Trimethylacetate, 2,2-Dimethylpropionic Acid Chloromethyl Ester, Pivalic Acid Chloromethyl Ester, Trimethylacetic Acid Chloromethyl Ester

Formula: C₆H₁₁ClO₂
MW: 150.6 g/mol
Boiling Pt: 147 °C
Density: 1.04 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00000884
CAS Number: 18997-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38f3ee60c5ee88d97dec88aa2be30387ea5292d9&vid=7be9b8f8140e43c2324dd36131cd5d2a28e7ba0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR