Propagyl amine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: Propargylamine hydrochloride, Prop-2-yn-1-amine hydrochloride

P0990-25G P0990-5G
TCP0990-25GEA 329.68 USD
TCP0990-25G TCP0990-5G
Propagyl amine hydrochloride ≥98.0%
Propagyl amine hydrochloride

CAS Number: 15430-52-1
MDL Number: MFCD00012907
Molecular Formula: C3H5N
Molecular Weight: 91.54
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White
Melting point (°C): 180


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Propynylamine Hydrochloride, 3-Amino-1-propyne Hydrochloride

Formula: C₃H₆ClN
MW: 91.54
Melting Pt: 180 °C
MDL Number: MFCD00012907
CAS Number: 15430-52-1
UN: 2811
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e7b93e11b01c5d9aae6246da87ce6e678fd3e87&vid=3e7b93e11b01c5d9aae6246da87ce6e678fd3e87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR