4-(1-Pyrrolidinyl)pyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Pyrrolidinopyridine

P0939-25G P0939-5G
TCP0939-25GEA 120 USD
TCP0939-25G TCP0939-5G
4-(1-Pyrrolidinyl)pyridine ≥98.0% (by titrimetric analysis)
4-(1-Pyrrolidinyl)pyridine

CAS Number: 2456-81-7
MDL Number: MFCD00006415
Molecular Formula: C9H12N2
Molecular Weight: 148.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 170
Melting point (°C): 58
Flash Point (°C): 160


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: 1-(4-Pyridyl)pyrrolidine

Formula: C₉H₁₂N₂
MW: 148.21 g/mol
Boiling Pt: 170 °C
Melting Pt: 58 °C
Flash Pt: 160 °C
MDL Number: MFCD00006415
CAS Number: 2456-81-7
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1dedf2017fc69104e15f6b0c140901dae19c6ef&vid=d1dedf2017fc69104e15f6b0c140901dae19c6ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR