Pentafluorobenzenesulfonyl chloride ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 832-53-1
MDL Number: MFCD00007427
Molecular Formula: C6ClF5O2S
Molecular Weight: 266.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 210
Flash Point (°C): 90
Specific Gravity (20/20): 1.81
Formula:
C₆ClF₅O₂S Boiling Pt: 210 °C Density: 1.81 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00007427 CAS Number: 832-53-1 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...