Pentafluoropyridine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,3,4,5,6-Perfluoropyridine, 2,3,4,5,6-Pentafluoropyridine

P0926-25G P0926-5G
TCP0926-25GEA 211 USD
TCP0926-25G TCP0926-5G
Pentafluoropyridine ≥98.0% (by GC)
Pentafluoropyridine

CAS Number: 700-16-3
MDL Number: MFCD00006225
Molecular Formula: C5F5N
Molecular Weight: 169.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 83
Flash Point (°C): 25
Specific Gravity (20/20): 1.62


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluoropyridine

Formula: C₅F₅N
MW: 169.05 g/mol
Boiling Pt: 83 °C
Density: 1.62 (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00006225
CAS Number: 700-16-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=133c313fe9c42ef327f060eb57fbe6643378784d&vid=133c313fe9c42ef327f060eb57fbe6643378784d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR