2,3,4,5,6-Pentafluorostyrene ≥98.0% (by GC) stabilized

Supplier: TCI America

P0862-25G P0862-5G
TCP0862-25GEA 240.41 USD
TCP0862-25G TCP0862-5G
2,3,4,5,6-Pentafluorostyrene ≥98.0% (by GC) stabilized
2,3,4,5,6-Pentafluorostyrene

(stabilized with TBC)
CAS Number: 653-34-9
MDL Number: MFCD00000300
Molecular Formula: C8H3F5
Molecular Weight: 194.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 34
Specific Gravity (20/20): 1.43
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,3,4,5,6-Pentafluorovinylbenzene

Formula: C₈H₃F₅
MW: 194.1 g/mol
Boiling Pt: 140 °C
Density: 1.43 (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00000300
CAS Number: 653-34-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e6fc5b3045e544bd84bb87404fdf6c32d079d11&vid=5e6fc5b3045e544bd84bb87404fdf6c32d079d11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR