2-Phenylpropionitrile ≥90.0% (by GC)

Supplier: TCI America

Synonyms: alpha-Methylphenylacetonitrile

P0860-10ML P0860-1ML
TCP0860-10MLEA 160 USD
TCP0860-10ML TCP0860-1ML
2-Phenylpropionitrile ≥90.0% (by GC)
2-Phenylpropionitrile

CAS Number: 1823-91-2
MDL Number: MFCD00001871
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 94
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: alpha-Methylphenylacetonitrile

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 230 °C
Density: 0.99 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00001871
CAS Number: 1823-91-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e1fe24973548c8674b6921df12507e6a7e708e9&vid=7e1fe24973548c8674b6921df12507e6a7e708e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR