Pentafluorobenzoyl chloride ≥98.0%

Supplier: TCI America

Synonyms: 2,3,4,5,6-Pentafluorobenzoyl chloride, Pentafluorobenzoylchloride, PFBC

P0807-25G P0807-5G
TCP0807-25GEA 229 USD
TCP0807-25G TCP0807-5G
Pentafluorobenzoyl chloride ≥98.0%
Pentafluorobenzoyl chloride

CAS Number: 2251-50-5
MDL Number: MFCD00000657
Molecular Formula: C7ClF5O
Molecular Weight: 230.52
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 159
Specific Gravity (20/20): 1.67


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Other Fluorinated Group Introducing Agents, Analytical Science, Acylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: Perfluorobenzoyl Chloride

Formula: C₇ClF₅O
Boiling Pt: 159 °C
Density: 1.67 (20 °C)
MDL Number: MFCD00000657
CAS Number: 2251-50-5
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb608a7184819bc98159ba236231cd2bb4d6ed18&vid=eb608a7184819bc98159ba236231cd2bb4d6ed18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR