(S)-(-)-1-Phenylethanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (S)-(-)-α-Methylbenzyl alcohol, (S)-(-)-sec-Phenethyl alcohol, (S)-1-Phenylethanol

P0796-1G P0796-25G P0796-5G
TCP0796-1GEA 75.3 USD
TCP0796-1G TCP0796-25G TCP0796-5G
(S)-(-)-1-Phenylethanol ≥98.0% (by GC)
(S)-(-)-1-Phenylethanol

CAS Number: 1445-91-6
MDL Number: MFCD00064264
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 85
Specific Gravity (20/20): 1.01
Specific rotation [a]20/D: -43 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (S)-(-)-alpha-Methylbenzyl Alcohol, (S)-(-)-1-Phenylethanol, (S)-(-)-sec-Phenethyl Alcohol

Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 98 °C
Density: 1.01 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00064264
CAS Number: 1445-91-6
UN: 2937
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7937c214aa986c7cfebbcbed8e78d2ec8a0a2ae8&vid=7937c214aa986c7cfebbcbed8e78d2ec8a0a2ae8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR