(R)-(+)-1-Phenylethanol ≥98.0%

Supplier: TCI America

Synonyms: (R)-1-Phenylethanol, (R)-(+)-α-Methylbenzyl alcohol, (R)-(+)-sec-Phenethyl alcohol

P0795-1G P0795-25G P0795-5G
TCP0795-1GEA 62 USD
TCP0795-1G TCP0795-25G TCP0795-5G
(R)-(+)-1-Phenylethanol ≥98.0%
(R)-(+)-1-Phenylethanol

CAS Number: 1517-69-7
MDL Number: MFCD00064263
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 89
Melting point (°C): 11
Flash Point (°C): 86
Specific Gravity (20/20): 1.01


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (R)-(+)-alpha-Methylbenzyl Alcohol, (R)-(+)-1-Phenylethanol, (R)-(+)-sec-Phenethyl Alcohol

Formula: C₈H₁₀O
Boiling Pt: 89 °C
Melting Pt: 11 °C
Density: 1.01 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00064263
CAS Number: 1517-69-7
UN: 2937
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1193df44239755dc9a822136ae1701f3307f5aa&vid=e1193df44239755dc9a822136ae1701f3307f5aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR