Dichloro(methyl)(phenyl)silane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methylphenyldichlorosilane

P0790-250G P0790-25G
TCP0790-250GEA 335 USD
TCP0790-250G TCP0790-25G
Dichloro(methyl)(phenyl)silane ≥98.0% (by GC)
Dichloro(methyl)(phenyl)silane

CAS Number: 149-74-6
MDL Number: MFCD00000490
Molecular Formula: C7H8Cl2Si
Molecular Weight: 191.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 82
Specific Gravity (20/20): 1.17

Formula: C₇H₈Cl₂Si
MW: 191.13 g/mol
Boiling Pt: 205 °C
Density: 1.17 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00000490
CAS Number: 149-74-6
UN: 2437
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b289c5b7768fd71344701714a3b7bb1eaf6d552f&vid=b289c5b7768fd71344701714a3b7bb1eaf6d552f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR